Learn More
Selleck Chemical LLC Aciclovir 50mg 59277-89-3 Acyclovir, Acycloguanosine, Zovirax
SDP
Supplier:  Selleck Chemical LLC S180750MG
Encompass_Preferred
Acyclovir (Acyclovir, Acycloguanosine, Zovirax) is a synthetic nucleoside analogue active against herpesviruses. Acyclovir induces cell cycle perturbation and apoptosis in Jurkat leukemia cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1652