Learn More
Electron Microscopy Sciences Acid Alcohol 1% 500 ML
SDP
Catalog No. 5031898
Encompass_Preferred
Used in Ehrlich's Hematoxylin , Mallory (1938); Ehrlich (1886), Nuclei.
Catalog No. 50-318-98 Supplier Electron Microscopy Sciences Supplier No. 2675302
Used in Ehrlich's Hematoxylin , Mallory (1938); Ehrlich (1886), Nuclei.