Learn More
Electron Microscopy Sciences Acid Alcohol, 1% HCl/70% Alcohol 1 LT
SDP
Supplier:  Electron Microscopy Sciences 2607201
Encompass_Preferred
Acidified alcohol solution is used for the differentiation of regressive stains. The solutions are freshly prepared upon receipt of order to ensure maximum shelf life.
Catalog No. 50-244-40