Learn More
Volu Sol Acid Alcohol Decolorizer, 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VAA128
Encompass_Preferred
Decolorizer, for use in staining Acid Fast Bacteria in the Kinyoun, Modified Kinyoun, or Ziehl-Neelsen staining protocols
Catalog No. 50-420-813