Learn More
Volu Sol Acid Alcohol Decolorizer, 32 oz / 1 L, Volu-Sol™
Supplier: Volu Sol VAA032
Decolorizer, for use in staining Acid Fast Bacteria in the Kinyoun, Modified Kinyoun, or Ziehl-Neelsen staining protocols