Learn More
Electron Microscopy Sciences Acid Alcohol, 0.5% HCl/70% Alcohol 1 LT
SDP
Catalog No. 5024442
Encompass_Preferred
Acidified alcohol solution is used for the differentiation of regressive stains. The solutions are freshly prepared upon receipt of order to ensure maximum shelf life.
Catalog No. 50-244-42 Supplier Electron Microscopy Sciences Supplier No. 2607101
Acidified alcohol solution is used for the differentiation of regressive stains. The solutions are freshly prepared upon receipt of order to ensure maximum shelf life.