Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Alcohol, ENG Scientific

Catalog No. ES6961
Change view
Click to view available options
Quantity:
2 x 250 mL
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6961 1 gal.
ES6960 2 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES6961 Supplier Eng Scientific Inc Supplier No. 6961
Add to Cart
Add to Cart

ENG Scientific Acid Alcohol uses a phenolated fluorescent dye in place of carbol fuchsin.

  • Flourescent Stain Kit for acid-fast bacteria
  • Acid Alcohol, Solution II

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 702
ChEBI CHEBI:16236

Specifications

Packaging Bottle
Quantity 1 gal.
Chemical Name or Material Acid Alcohol

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.