Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Anthracene Brown PG, TCI America™
SDP

Catalog No. A066425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A066425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A066425G Supplier TCI America Supplier No. A066425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6369-32-0
Molecular Formula C23H15N3Na2O6S
Molecular Weight (g/mol) 507.43
MDL Number MFCD00059537
InChI Key GDVUVRLDXFWCSM-CXJWIYSGSA-L
Synonym Chrome Brown PG
PubChem CID 131864341
IUPAC Name disodium 2-{2-[(2E)-1-oxo-7-(phenylamino)-3-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1N\N=C1/C(=O)C2=CC(NC3=CC=CC=C3)=CC=C2C=C1S([O-])(=O)=O

Specifications

Color Black-Purple
Quantity 25 g
Formula Weight 507.43
Physical Form Crystalline Powder
Chemical Name or Material Acid Anthracene Brown PG
EINECSNumber (5)-2107
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.