Learn More
Ethos Biosciences Trichloroacetic Acid 10%, 4oz, Ethos
Catalog No. 5000574296
Encompass_Preferred
Trichloroacetic Acid 10% is a ready-to-use aqueous solution suitable for general laboratory and analytical applications. Commonly used for protein precipitation, reagent preparation, and chemical analysis, it supports routine laboratory workflows requiring a dilute trichloroacetic acid solution. The clear, colorless solution provides consistent concentration for reliable and reproducible results. It is also appropriate for cleaning, sample preparation, and other general-purpose laboratory procedures.
Catalog No. 50-005-74296 Supplier Ethos Biosciences Supplier No. 350104
Trichloroacetic Acid 10% is a ready-to-use aqueous solution suitable for general laboratory and analytical applications. Commonly used for protein precipitation, reagent preparation, and chemical analysis, it supports routine laboratory workflows requiring a dilute trichloroacetic acid solution. The clear, colorless solution provides consistent concentration for reliable and reproducible results. It is also appropriate for cleaning, sample preparation, and other general-purpose laboratory procedures.