Learn More
Ethos Biosciences Hydrochloric Acid, Concentrated, 16oz, Ethos
Catalog No. 5000574233
Encompass_Preferred
Hydrochloric Acid, Concentrated is a high-strength mineral acid suitable for general laboratory, industrial, and chemical processing applications. Commonly used for pH adjustment, titrations, reagent preparation, and chemical synthesis, it delivers consistent performance for a wide range of analytical and production tasks. This clear, colorless to slightly yellow solution provides reliable concentration for reproducible results in routine and specialized procedures. It is also appropriate for cleaning, metal treatment, and general chemical processing. Supplied in chemically compatible containers, Concentrated Hydrochloric Acid is suitable for research, pharmaceutical, educational, and industrial laboratory environments.
Catalog No. 50-005-74233 Supplier Ethos Biosciences Supplier No. 340016
Hydrochloric Acid, Concentrated is a high-strength mineral acid suitable for general laboratory, industrial, and chemical processing applications. Commonly used for pH adjustment, titrations, reagent preparation, and chemical synthesis, it delivers consistent performance for a wide range of analytical and production tasks. This clear, colorless to slightly yellow solution provides reliable concentration for reproducible results in routine and specialized procedures. It is also appropriate for cleaning, metal treatment, and general chemical processing. Supplied in chemically compatible containers, Concentrated Hydrochloric Acid is suitable for research, pharmaceutical, educational, and industrial laboratory environments.