Learn More
Ethos Biosciences Hydrochloric Acid 1N, 16oz, Ethos
Catalog No. 5000574197
Encompass_Preferred
Hydrochloric Acid 1N is a standardized acid solution suitable for general laboratory and industrial applications. Commonly used for pH adjustment, titrations, reagent preparation, and analytical procedures, it supports consistent and reliable results across routine workflows. The clear, colorless solution provides dependable concentration for reproducible performance. It is also appropriate for cleaning, chemical processing, and other general-purpose laboratory tasks. Supplied in chemically compatible containers, Hydrochloric Acid 1N is suitable for research, pharmaceutical, educational, and industrial laboratory environments.
Catalog No. 50-005-74197 Supplier Ethos Biosciences Supplier No. 339616
Hydrochloric Acid 1N is a standardized acid solution suitable for general laboratory and industrial applications. Commonly used for pH adjustment, titrations, reagent preparation, and analytical procedures, it supports consistent and reliable results across routine workflows. The clear, colorless solution provides dependable concentration for reproducible performance. It is also appropriate for cleaning, chemical processing, and other general-purpose laboratory tasks. Supplied in chemically compatible containers, Hydrochloric Acid 1N is suitable for research, pharmaceutical, educational, and industrial laboratory environments.