Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Acid and Base Neutralizing Polymers

Catalog No. 19129310 Shop All Brady Corporation Products
Change view
Click to view available options
Type:
Base Neutralizing Polymer
Color:
Turns orange
Turns purple
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Color
19-129-310 Base Neutralizing Polymer Turns orange
19-129-309 - Turns purple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-129-310 Supplier Brady™ Supplier No. SPCBASE
  • $79.00
    $65.65 / Each of 1

    Save $13.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Neutralizing Polymers are used to solidify a wide range of acid and base spills

  • Provides safe spill control
  • Polymers neutralize and change color when neutralization has taken place
    • Acid neutralizer turns purple
    • Base neutralizer turns orange
  • Non-toxic, non-corrosive and non-hazardous

Specifications

Color Turns orange
Capacity (English) 2.6 qt.
Type Base Neutralizing Polymer
For Use With (Application) Base Chemical Spills
Packaging Quantity 1 Ea.
Dimensions (L x W) 12.5 x 9.5 in.
Length (English) 12.5 in.
Width (English) 12.5 in.
Height (English) 9.5 in
Depth (English) 12.5 in
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.