Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Black 1 97.0+%, TCI America™
SDP

Catalog No. A0586100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0586100G 100 g
A058625G 25 g
A0586500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0586100G Supplier TCI America Supplier No. A0586100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
MDL Number MFCD00004017
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym amido black 10b, acid black 1, buffalo black nbr, naphthol blue black, parent, aniline blue black, tech, nigrosine water soluble, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo naphthalene-2,7-disulphonic acid, sodium salt, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo-, sodium salt, 3-4-nitrophenylazo-4-amino-5-oxo-6-2-phenylhydrazono-5,6-dihydro-2,7-naphthalenedisulfonic acid disodium salt
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 100 g
Formula Weight 616.49
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Acid Black 1
EINECSNumber (5)-1875&(5)-5232
RTECSNumber QJ6196000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.