Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Blue 119, TCI America™
SDP

Catalog No. A05961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A05961G Supplier TCI America Supplier No. A05961G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1324-80-7
Molecular Formula C37H30N3NaO4S
Molecular Weight (g/mol) 635.714
MDL Number MFCD00083221
InChI Key BPCZPTGXEYXQRS-UHFFFAOYSA-M
Synonym Alkali Blue 6B
PubChem CID 71464047
IUPAC Name sodium;2-[4-[bis(4-anilinophenyl)-hydroxymethyl]anilino]benzenesulfonate
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)C(C3=CC=C(C=C3)NC4=CC=CC=C4)(C5=CC=C(C=C5)NC6=CC=CC=C6S(=O)(=O)[O-])O.[Na+]

Specifications

Color Blue/Red
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Acid Blue 119
EINECSNumber (5)-3116&(5)-3312
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.