Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acid-C1-PEG5-Boc | 2304558-22-1 | 97.0% | C17H32O9 | 25 MG
SDP

Catalog No. 5000357393
Encompass_Preferred

Acid-C1-PEG5-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It appears as a colorless to light yellow liquid.

  • Molecular weight: 380.43
  • Chemical formula: C17H32O9
  • Purity: 97.0%
  • Soluble in DMSO (125 mg/mL)
  • Store pure form at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-57393 Supplier Medchemexpress LLC Supplier No. 25MGHY140483
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acid-C1-PEG5-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It appears as a colorless to light yellow liquid.

  • Molecular weight: 380.43
  • Chemical formula: C17H32O9
  • Purity: 97.0%
  • Soluble in DMSO (125 mg/mL)
  • Store pure form at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.