Learn More
Medchemexpress LLC Acid-C1-PEG5-Boc | 2304558-22-1 | 97.0% | C17H32O9 | 50 MG
SDP
Catalog No. 5000357018
Encompass_Preferred
Acid-C1-PEG5-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
Catalog No. 50-003-57018 Supplier Medchemexpress LLC Supplier No. HY14048350MG
Acid-C1-PEG5-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins