Learn More
Medchemexpress LLC Acid-C2-PEG4-C2-NHS ester | 1343476-41-4 | 98.0% | 25 MG
SDP
Catalog No. 5000339507
Encompass_Preferred
Acid-C2-PEG4-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase and the other for the target protein
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-39507 Supplier Medchemexpress LLC Supplier No. 25MGHY130436
Acid-C2-PEG4-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase and the other for the target protein
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins