Learn More
Medchemexpress LLC Acid-C3-SSPy | 250266-79-6 | 229.32 g/mol | C9H11NO2S2 | 5 MG
SDP
Catalog No. 5000203984
Encompass_Preferred
Acid-C3-SSPy is a cleavable DBA-DM4 linker intended for use as a linker intermediate in antibody-drug conjugate (ADC) synthesis. It has molecular formula C9H11NO2S2 and molecular weight 229.32 g/mol, and is supplied in small-scale quantities for research applications.
  • Cleavable disulfide-based linker suitable for ADC conjugation
  • Compact molecular weight for facile conjugation chemistry
  • Available in small quantities for research-scale synthesis
  • Suitable for use as a linker intermediate in ADC construction
Catalog No. 50-002-03984 Supplier Medchemexpress LLC Supplier No. HY1415975MG
Add to Cart
Add to Cart
Acid-C3-SSPy is a cleavable DBA-DM4 linker intended for use as a linker intermediate in antibody-drug conjugate (ADC) synthesis. It has molecular formula C9H11NO2S2 and molecular weight 229.32 g/mol, and is supplied in small-scale quantities for research applications.
  • Cleavable disulfide-based linker suitable for ADC conjugation
  • Compact molecular weight for facile conjugation chemistry
  • Available in small quantities for research-scale synthesis
  • Suitable for use as a linker intermediate in ADC construction