Learn More
Medchemexpress LLC HY-141866 5mg , Acid Ceramidase-IN-1 CAS:2415225-30-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1418665MG
Encompass_Preferred
HY-141866 5mg Acid Ceramidase-IN-1 CAS: 2415225-30-6 Acid Ceramidase-IN-1 is a potent and oral bioavailable acid ceramidase (AC, ASAH-1) inhibitor (hAC IC50=0.166 μM). Acid Ceramidase-IN-1 has excellent brain penetration in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1459