Learn More
Medchemexpress LLC Acid Ceramidase-IN-2 | 1005497-03-9 | C30H47NO6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1533961MG
Encompass_Preferred
Acid Ceramidase-IN-2 is an acid ceramidase inhibitor with potential antiproliferative and cytostatic activities. Human acid ceramidase is overexpressed in prostate cancer cells, suggesting a potential anti-tumor effect. Its hydrolysis can be inhibited in vitro by 3 α-ketoamides: GT85, GT98, and GT99.
  • Potential antiproliferative and cytostatic activities
  • Inhibits human acid ceramidase, which is overexpressed in prostate cancer cells
  • Hydrolysis inhibited by 3 α-ketoamides: GT85, GT98, and GT99
Catalog No. 50-003-73541