Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Dichromate Cleaning Solution, Reagents

Catalog No. p-200021753
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-061 1 L
01-336-060 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-061 Supplier Reagents Holdings Llc Supplier No. CA0134001C1
Only null left
Add to Cart
Add to Cart

Acid Dichromate Cleaning Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color-coded No
Form Liquid
pH <1
Sterility Non-sterile
Autoclavable Not Autoclavable
Boiling Point 300°C
Color Brown
Format Liquid
Quantity 1 L
Solubility Completely miscible
Volume (Metric) 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.