Learn More
Scytek Laboratories Acid Fast Bacteria (AFB) Stain Kit: Small
SDP
Catalog No. NC1969820
Encompass_Preferred
The Acid Fast Bacteria (AFB) Stain Kit is intended for use in the histological visualization of Acid Fast Bacteria and Tubercle Bacilli. This kit is a rapid 15-minute procedure. The lipoid capsule of the acid-fast organism takes up carbol fuchsin and resists decolorization.Acid Fast Organisms: Bright RedBackground: Light GreenKit contents: CFZ030, Carbol Fuchsin Solution, 30 ml. AAL030, Acid Alcohol Solution (0.5%), 2 x 30 ml.LGA030, Light Green Solution, 30 ml.
This item is not returnable. View return policy
Catalog No. NC1969820 Supplier Scytek Laboratories Supplier No. FAB2
The Acid Fast Bacteria (AFB) Stain Kit is intended for use in the histological visualization of Acid Fast Bacteria and Tubercle Bacilli. This kit is a rapid 15-minute procedure. The lipoid capsule of the acid-fast organism takes up carbol fuchsin and resists decolorization.Acid Fast Organisms: Bright RedBackground: Light GreenKit contents: CFZ030, Carbol Fuchsin Solution, 30 ml. AAL030, Acid Alcohol Solution (0.5%), 2 x 30 ml.LGA030, Light Green Solution, 30 ml.