Learn More
StatLab Acid Fast Control Slides
Description
- AFB control slides simulate clinical sputum smears with appropriate background material in both positive and negative wells
- Positive well: Mycobacteria tuberculosis
- Negative well: Escherichia coli
- 50 slides/box
Specifications
Specifications
| Product Type | Control Slide |
| Quantity | 50 Slides/Box |