Learn More
Electron Microscopy Sciences Acid Fuchsin 0.5% Aqueous 500 ML
SDP
Catalog No. 5031925
Encompass_Preferred
Acid Fuchsin Aqueous solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results.
Catalog No. 50-319-25 Supplier Electron Microscopy Sciences Supplier No. 2602005
Acid Fuchsin Aqueous solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results.