Learn More
Electron Microscopy Sciences Acid-Fuchsin Aniline Solution 500 ML
SDP
Catalog No. 5031737
Encompass_Preferred
Used in Laqueur's Method for Alcoholic Hyalin for mallory bodies, erythrocytes, bile pigment and proteinaceous, material in liver.
Laqueur, G.I., Amer. J. Clin. Path., 20:689-690, 1950, Luna, L.G. (ed), Manual of Hitopathologic Staining Method of the AFIP, 3rd ed, McGraw-Hill, NY, c. 1968, p 160
Lillie, R.D., Fullmer, H.M., Histopathologic Technic and Practical Histochemistry 4th ed., cGraw-Hill, NY, 1976, p 670-671
Clark, G., (ed) Staining Procedures, 3rd ed. WilliamS & Wilkins, Baltimore, p 51, c. 1973
Catalog No. 50-317-37 Supplier Electron Microscopy Sciences Supplier No. 2630602
Used in Laqueur's Method for Alcoholic Hyalin for mallory bodies, erythrocytes, bile pigment and proteinaceous, material in liver.
Laqueur, G.I., Amer. J. Clin. Path., 20:689-690, 1950, Luna, L.G. (ed), Manual of Hitopathologic Staining Method of the AFIP, 3rd ed, McGraw-Hill, NY, c. 1968, p 160
Lillie, R.D., Fullmer, H.M., Histopathologic Technic and Practical Histochemistry 4th ed., cGraw-Hill, NY, 1976, p 670-671
Clark, G., (ed) Staining Procedures, 3rd ed. WilliamS & Wilkins, Baltimore, p 51, c. 1973