Learn More
Alkali Scientific Acid Fuchsin Calcium Salt, 100 Grams
SDP
Catalog No. 5000660617
Encompass_Preferred
Acid Fuchsin Calcium Salt, 100 Grams, is a biological stain used in histology, microbiology, and other laboratory procedures. This reagent is commonly used to color cellular components, such as cytoplasm and nucleic acids, providing contrast during microscopic examination. Acid fuchsin is also utilized in the detection of specific cellular structures, making it valuable for diagnostic and research applications. This calcium salt form enhances the stability and solubility of the dye, ensuring consistent results in staining protocols. The 100g quantity is ideal for laboratories performing routine histological staining, microbiological culture work, or other cellular analyses.
Catalog No. 50-006-60617 Supplier Alkali Scientific Supplier No. ACI001CP
Acid Fuchsin Calcium Salt, 100 Grams, is a biological stain used in histology, microbiology, and other laboratory procedures. This reagent is commonly used to color cellular components, such as cytoplasm and nucleic acids, providing contrast during microscopic examination. Acid fuchsin is also utilized in the detection of specific cellular structures, making it valuable for diagnostic and research applications. This calcium salt form enhances the stability and solubility of the dye, ensuring consistent results in staining protocols. The 100g quantity is ideal for laboratories performing routine histological staining, microbiological culture work, or other cellular analyses.