Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Fuchsin Calcium Salt, Sigma-Aldrich

Catalog No. 01259019 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-019 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-019 Supplier MilliporeSigma Supelco Supplier No. 857408100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acid fuchsine (C. I. 42685) is a sulfonated triaminotriphenylmethane. It is also known as acid magenta. At pH 12-14, acid fuchsine changes from red to colorless.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-10-1
Molecular Weight (g/mol) 579.64 g/mol

Specifications

Concentration ≥60%
Color Red
Quantity 100 g
Grade Certified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.