Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Acid Fuchsin, Certified, C.I. DcR-8 25 GR
SDP

Catalog No. 50980151
Encompass_Preferred

(Rubin S, Fuchsin S, Fuchsin Acid, Acid Magenta, Acid Violet 19)
CAS #3244-88-0 C₂₀H₁₇N₃Na₂O₉S₃ F.W. 585.54
Solubility: 0.053% Water; 1.21% Alcohol

Acid Fuchsin (Rubin S, Fuchsin S, Fuchsin Acid, Acid Magenta, Acid Violet 19)

Catalog No. 50-980-151 Supplier Electron Microscopy Sciences Supplier No. 10035
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rubin S, Fuchsin S, Fuchsin Acid, Acid Magenta, Acid Violet 19)
CAS #3244-88-0 C₂₀H₁₇N₃Na₂O₉S₃ F.W. 585.54
Solubility: 0.053% Water; 1.21% Alcohol

Acid Fuchsin (Rubin S, Fuchsin S, Fuchsin Acid, Acid Magenta, Acid Violet 19)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.