Learn More
Dentec Safety™ Acid Gas and Chlorine Dioxide Cartridge
SDP
Catalog No. 19222509 Shop All Dentec Safety Specialists Inc. Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100/Box
6/Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-222-509 6/Box
19-222-510 100/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 19-222-509 missing translation for 'mfr' Dentec Safety™ missing translation for 'supplierNo' 158T21
Add to Cart
Add to Cart
Dentec Safety™ Acid Gas and Chlorine Dioxide Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity.
  • Offset connector allows the user to rotate the cartridge into 3 different positions improves visibility and allows face shields or welding helmets to be worn without interference
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
Quantity 6/Box