Learn More
Dentec Safety™ Acid Gas and Organic Vapor Cartridge
SDP
Catalog No. 19222514 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Quantity:
100/Box
6/Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-222-514 100/Box
19-222-513 6/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-514 Supplier Dentec Safety™ Supplier No. 158T26B100
Add to Cart
Add to Cart
Dentec Safety™ Acid Gas and Organic Vapor Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity
  • Offset connector allows the user to rotate the cartridge into 3 different positions improves visibility and allows face shields or welding helmets to be worn without interference
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
Quantity 100/Box