Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Green 3, TCI America™
SDP

Catalog No. G017625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G017625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G017625G Supplier TCI America Supplier No. G017625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4680-78-8
Molecular Formula C37H35N2NaO6S2
Molecular Weight (g/mol) 690.805
MDL Number MFCD00012122
InChI Key XKTMIJODWOEBKO-UHFFFAOYSA-M
Synonym Acid Green B, Guinea Green B, Pontacyl Green B
PubChem CID 20803
IUPAC Name sodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-phenylmethyl]anilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5.[Na+]

Specifications

Melting Point 255°C
Color Purple/Red
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 690.80
Chemical Name or Material Acid Green 3
EINECSNumber (5)-1734
RTECSNumber BQ4375000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.