Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Green 5 MP Biomedicals

Catalog No. ICN15264125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15264125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15264125 Supplier MP Biomedicals, Inc Supplier No. 0215264125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Histological and cytological stain used for plasma counterstaining in laboratory procedures.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5141-20-8
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
InChI Key DGOBMKYRQHEFGQ-UHFFFAOYSA-L
Synonym Acid Green 5, FD & C Green No 2
PubChem CID 21223
ChEBI CHEBI:87065
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Green
Quantity 25 g
Formula Weight 792.8
Grade Histological
Chemical Name or Material Acid Green 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.