Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Green 9, TCI America™
SDP

Catalog No. B079125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B079125G Supplier TCI America Supplier No. B079125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic dye powder for use in research applications.

Chemical Identifiers

CAS 4857-81-2
Molecular Formula C37H34ClN2NaO6S2
Molecular Weight (g/mol) 725.247
MDL Number MFCD00050440
InChI Key SHBDDIJUSNNBLQ-UHFFFAOYSA-M
Synonym Patent Green, Alizarin Green, Brilliant Acid Green, Brilliant Milling Green
PubChem CID 25021982
IUPAC Name sodium;3-[[4-[(2-chlorophenyl)-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5Cl.[Na+]

Specifications

Color Black/Purple
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 725.25
Chemical Name or Material Acid Green 9
EINECSNumber (5)-1736
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.