Learn More
NEOGEN Acid Hydrolysed Casein, 10 kg
SDP
Catalog No. 502015824
Encompass_Preferred
Acid Hydrolysed Casein is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.A soluble protein hydrolysate obtained by digesting casein with hot acid. It is almost free from growth factors, vitamins and antagonists, and these qualities make it suitable for use as a protein source in media for antibiotic and vitamin assays.For more information visit https://www.neogen.com/solutions/Microbiology/acid-hydrolysed-casein
Catalog No. 50-201-5824 Supplier NEOGEN Supplier No. 700004683
Acid Hydrolysed Casein is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.A soluble protein hydrolysate obtained by digesting casein with hot acid. It is almost free from growth factors, vitamins and antagonists, and these qualities make it suitable for use as a protein source in media for antibiotic and vitamin assays.For more information visit https://www.neogen.com/solutions/Microbiology/acid-hydrolysed-casein