Learn More
Crystal Chem Inc ALS (Acid Labile Subunit) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80584
Encompass
The Acid Labile Subunit ELISA is used to quantify levels of human acid labile subunit (ALS) in serum and plasma using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. Dynamic range of 1.5 - 40 mU/mL. High sensitivity of 0.23 mU/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7750