Learn More
Medchemexpress LLC Acid-PEG1-C2-Boc | 2086689-08-7 | C10H18O5 | 100 MG
SDP
Catalog No. 5000353041
Encompass_Preferred
Acid-PEG1-C2-Boc is a PEG-based PROTAC linker designed for research purposes. It can be used in the synthesis of PROTACs, which are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrade target proteins
Catalog No. 50-003-53041 Supplier Medchemexpress LLC Supplier No. HY140479100MG
Acid-PEG1-C2-Boc is a PEG-based PROTAC linker designed for research purposes. It can be used in the synthesis of PROTACs, which are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrade target proteins