Learn More
Medchemexpress LLC Acid-PEG2-SS-PEG2-acid | 1807539-10-1 | 95.0% | 386.48 g/mol | C14H26O8S2 | 10 MG
SDP
Catalog No. 5000203775
Encompass_Preferred
Acid-PEG2-SS-PEG2-acid is a cleavable four-unit polyethylene glycol (PEG) linker containing a central disulfide bond and terminal carboxylic acid groups. It serves as a hydrophilic spacer and a reducible linkage for antibody-drug conjugate (ADC) synthesis and bioconjugation applications. The product is supplied for research use only and is not for human use.
  • Cleavable disulfide bond enabling payload release in reducing environments
  • Four-unit PEG spacer increases hydrophilicity and solubility
  • Terminal carboxylic acids enable common conjugation chemistries
  • Available in multiple pack sizes, including 10 mg
  • Typical purity around 95.0%
Catalog No. 50-002-03775 Supplier Medchemexpress LLC Supplier No. HY14011210MG
Acid-PEG2-SS-PEG2-acid is a cleavable four-unit polyethylene glycol (PEG) linker containing a central disulfide bond and terminal carboxylic acid groups. It serves as a hydrophilic spacer and a reducible linkage for antibody-drug conjugate (ADC) synthesis and bioconjugation applications. The product is supplied for research use only and is not for human use.
  • Cleavable disulfide bond enabling payload release in reducing environments
  • Four-unit PEG spacer increases hydrophilicity and solubility
  • Terminal carboxylic acids enable common conjugation chemistries
  • Available in multiple pack sizes, including 10 mg
  • Typical purity around 95.0%