Learn More
Medchemexpress LLC Acid-PEG2-SS-PEG2-acid | 1807539-10-1 | 95.0% | 386.48 g/mol | C14H26O8S2 | 5 MG
SDP
Catalog No. 5000202644
Encompass_Preferred
Acid-PEG2-SS-PEG2-acid is a cleavable 4-unit polyethylene glycol (PEG) linker containing a central disulfide bond and terminal carboxylic acids, designed for use in antibody-drug conjugate (ADC) synthesis and bioconjugation. Supplied as a white to off-white solid for research use only.
  • Cleavable disulfide bond enables intracellular release under reducing conditions.
  • Terminal carboxyl groups allow amide coupling and conjugation chemistry.
  • Four-unit PEG spacer improves solubility and reduces steric hindrance.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance with recommended cold storage.
Catalog No. 50-002-02644 Supplier Medchemexpress LLC Supplier No. HY1401125MG
Acid-PEG2-SS-PEG2-acid is a cleavable 4-unit polyethylene glycol (PEG) linker containing a central disulfide bond and terminal carboxylic acids, designed for use in antibody-drug conjugate (ADC) synthesis and bioconjugation. Supplied as a white to off-white solid for research use only.
  • Cleavable disulfide bond enables intracellular release under reducing conditions.
  • Terminal carboxyl groups allow amide coupling and conjugation chemistry.
  • Four-unit PEG spacer improves solubility and reduces steric hindrance.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance with recommended cold storage.