Learn More
Medchemexpress LLC Acid-PEG3-C2-Boc | 1807539-06-5 | ≥97.0% | 100 MG
SDP
Catalog No. 5000339973
Encompass_Preferred
Acid-PEG3-C2-Boc is a PEG- and Alkyl/ether-based PROTAC linker. It can be used in the synthesis of PROTACs for the degradation of EGFR and inhibition of mTOR. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg- and alkyl/ether-based protac linker
  • Can be used in the synthesis of protacs
  • Potential for degradation of egfr
  • Potential for inhibition of mtor
Catalog No. 50-003-39973 Supplier Medchemexpress LLC Supplier No. HY130549100MG
Acid-PEG3-C2-Boc is a PEG- and Alkyl/ether-based PROTAC linker. It can be used in the synthesis of PROTACs for the degradation of EGFR and inhibition of mTOR. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg- and alkyl/ether-based protac linker
  • Can be used in the synthesis of protacs
  • Potential for degradation of egfr
  • Potential for inhibition of mtor