Learn More
Medchemexpress LLC Acid-PEG3-SS-PEG3-acid | 2055014-98-5 | 474.59 g·mol⁻¹ | C18H34O10S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1401135MG
Encompass_Preferred
This cleavable six-unit polyethylene glycol (PEG) linker is designed for the synthesis of antibody-drug conjugates (ADCs). It contains a central disulfide bond flanked by PEG3 spacers and terminating carboxylic acid groups, enabling conjugation to payloads and reductive intracellular release.
  • cleavable disulfide (SS) bond for controlled payload release.
  • six-unit PEG scaffold increases solubility and flexibility.
  • terminal carboxylic acid groups enable standard conjugation chemistries.
  • suitable for antibody-drug conjugate (ADC) and bioconjugation workflows.
  • provided in multiple small pack sizes for research use.
Catalog No. 50-002-03690