Learn More
Medchemexpress LLC Acid-PEG3-SS-PEG3-acid | 2055014-98-5 | ≥95.0% | 474.59 | C18H34O10S2 | 50 MG
SDP
Catalog No. 5000202899
Encompass_Preferred
Acid-PEG3-SS-PEG3-acid is a cleavable six-unit polyethylene glycol (PEG) linker containing a central disulfide bond. It is designed for use in antibody-drug conjugate (ADC) synthesis and conjugation chemistry, providing a water-soluble spacer that enables intracellular payload release through disulfide cleavage.
  • Cleavable six-unit PEG linker with a disulfide (SS) bond.
  • Designed for ADC synthesis and conjugation chemistry.
  • Enables intracellular payload release via disulfide reduction.
  • PEG spacer improves aqueous solubility of conjugates.
  • Molecular weight 474.59; CAS 2055014-98-5; available in multiple pack sizes including 50 MG.
Catalog No. 50-002-02899 Supplier Medchemexpress LLC Supplier No. HY14011350MG
Acid-PEG3-SS-PEG3-acid is a cleavable six-unit polyethylene glycol (PEG) linker containing a central disulfide bond. It is designed for use in antibody-drug conjugate (ADC) synthesis and conjugation chemistry, providing a water-soluble spacer that enables intracellular payload release through disulfide cleavage.
  • Cleavable six-unit PEG linker with a disulfide (SS) bond.
  • Designed for ADC synthesis and conjugation chemistry.
  • Enables intracellular payload release via disulfide reduction.
  • PEG spacer improves aqueous solubility of conjugates.
  • Molecular weight 474.59; CAS 2055014-98-5; available in multiple pack sizes including 50 MG.