Learn More
Medchemexpress LLC Acid-PEG3-SS-PEG3-acid | 2055014-98-5 | 98.0% | 474.59 g/mol | C18H34O10S2 | 250 MG
SDP
Catalog No. 5000203689
Encompass_Preferred
Acid-PEG3-SS-PEG3-acid is a cleavable six-unit polyethylene glycol (PEG) linker comprising two PEG3 segments joined by a disulfide bond and terminating in carboxylic acid groups. It is formulated for use in antibody-drug conjugate (ADC) synthesis and related bioconjugation research, and is supplied in small research-scale packages.
  • Cleavable disulfide linker enabling intracellular payload release.
  • Six PEG units (two PEG3 segments) provide spacing and solubility.
  • Terminal carboxylic acids supply sites for conjugation chemistry.
  • Designed for ADC synthesis and related bioconjugation applications.
  • Available in multiple small-scale package sizes for research use.
Catalog No. 50-002-03689 Supplier Medchemexpress LLC Supplier No. HY140113250MG
Acid-PEG3-SS-PEG3-acid is a cleavable six-unit polyethylene glycol (PEG) linker comprising two PEG3 segments joined by a disulfide bond and terminating in carboxylic acid groups. It is formulated for use in antibody-drug conjugate (ADC) synthesis and related bioconjugation research, and is supplied in small research-scale packages.
  • Cleavable disulfide linker enabling intracellular payload release.
  • Six PEG units (two PEG3 segments) provide spacing and solubility.
  • Terminal carboxylic acids supply sites for conjugation chemistry.
  • Designed for ADC synthesis and related bioconjugation applications.
  • Available in multiple small-scale package sizes for research use.