Learn More
Medchemexpress LLC Acid-PEG3-SS-PEG3-acid | 2055014-98-5 | MFCD28950758 | 98.0% | 474.59 g·mol⁻¹ | C18H34O10S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140113100MG
Encompass_Preferred
Acid-PEG3-SS-PEG3-acid is a cleavable six-unit polyethylene glycol (PEG) linker that contains a central disulfide bond and terminal carboxylic acid groups. It is designed as a bioreducible spacer for use in the synthesis of antibody-drug conjugates (ADCs), providing hydrophilicity and a site for conjugation chemistry.
  • Cleavable disulfide (SS) linkage for intracellular payload release.
  • Two PEG3 units provide hydrophilicity and spacer length.
  • Terminal carboxylic acids enable common conjugation chemistries.
  • Molecular weight 474.59 g·mol⁻¹ and formula C18H34O10S2.
  • Available in small research quantities suitable for ADC development.
Catalog No. 50-002-03972