Learn More
Medchemexpress LLC Acid-PEG4-C2-Boc | 1835759-85-7 | 98.3% | 350.40 | 50 MG
SDP
Catalog No. 5000400960
Encompass_Preferred
Acid-PEG4-C2-Boc is a PEG- and Alkyl/ether-based PROTAC linker. It can be used in the synthesis of PROTACs for the inhibition of mTOR. This product is for research use only and is not sold to patients.
  • PEG- and alkyl/ether-based linker
  • Aids in the synthesis of PROTACs
  • Assists in the inhibition of mTOR
Catalog No. 50-004-00960 Supplier Medchemexpress LLC Supplier No. HY2316750MG
Acid-PEG4-C2-Boc is a PEG- and Alkyl/ether-based PROTAC linker. It can be used in the synthesis of PROTACs for the inhibition of mTOR. This product is for research use only and is not sold to patients.
  • PEG- and alkyl/ether-based linker
  • Aids in the synthesis of PROTACs
  • Assists in the inhibition of mTOR