Learn More
Medchemexpress LLC Acid-PEG4-S-S-PEG4-acid | 2055015-40-0 | 95% | 562.69 | 100 MG
SDP
Catalog No. 5000348004
Encompass_Preferred
Acid-PEG4-S-S-PEG4-acid is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This compound is intended for research applications only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Soluble at 10 mM in DMSO
  • For research use only
Catalog No. 50-003-48004 Supplier Medchemexpress LLC Supplier No. HY140114100MG
Acid-PEG4-S-S-PEG4-acid is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This compound is intended for research applications only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Soluble at 10 mM in DMSO
  • For research use only