Learn More
Pharmacal Research Laboratories Acid Power, 55 Gal
SDP
Catalog No. NC0926413
Encompass
Acid-Power is a phosphoric acid washing detergent formulated with inhibitors that minimize corrosion. This product contains non-ionic surfactants to increase the efficiency of removing urine scale deposits and organic soils.
Catalog No. NC0926413 Supplier Pharmacal Research Laboratories Supplier No. 05055
Acid-Power is a phosphoric acid washing detergent formulated with inhibitors that minimize corrosion. This product contains non-ionic surfactants to increase the efficiency of removing urine scale deposits and organic soils.