Learn More
Pharmacal Research Laboratories ACID POWR URINE SCAL REMOVE 30
SDP
Catalog No. NC1997448
Encompass
Acid-Power® 30 gallon drumAcid-Power® is a phosphoric acid washing detergent formulated with inhibitors that minimize corrosion. This product contains non-ionic surfactants to increase the efficiency of removing urine scale deposits and organic soils. Acid-Power is non-foaming and free rinsing. This compound is stable in wash temperatures in excess of 180o F and will not break down under these conditions
Catalog No. NC1997448 Supplier Pharmacal Research Laboratories Supplier No. 05030F
Acid-Power® 30 gallon drumAcid-Power® is a phosphoric acid washing detergent formulated with inhibitors that minimize corrosion. This product contains non-ionic surfactants to increase the efficiency of removing urine scale deposits and organic soils. Acid-Power is non-foaming and free rinsing. This compound is stable in wash temperatures in excess of 180o F and will not break down under these conditions