Learn More
Electron Microscopy Sciences Acid Pump/Reagent Dispenser
SDP
Catalog No. 501926887
Encompass_Preferred
Dispense dangerous liquids safely and easily withpout pouring reducing spills and breakage.Use light pressure on the relief valve button on top of the dispenser quickly stops the flow of liquid. Rapid but light finger tip squeezes on the small bottle will start fluid dispensing from the short tube. Fits many different sizes of containers including: 2.5 liter, 4 liter, 5 pint, 0.5 gallon, 1 gallon.
Includes two adapters for short neck bottles, dispenser body-threads with GPS No. 38-430 or 38-400 threads, short length of " diameter polyethylene tube for outlet, and Polyethylene tubing-" O.D.
Catalog No. 50-192-6887 Supplier Electron Microscopy Sciences Supplier No. 6429912
Dispense dangerous liquids safely and easily withpout pouring reducing spills and breakage.Use light pressure on the relief valve button on top of the dispenser quickly stops the flow of liquid. Rapid but light finger tip squeezes on the small bottle will start fluid dispensing from the short tube. Fits many different sizes of containers including: 2.5 liter, 4 liter, 5 pint, 0.5 gallon, 1 gallon.
Includes two adapters for short neck bottles, dispenser body-threads with GPS No. 38-430 or 38-400 threads, short length of " diameter polyethylene tube for outlet, and Polyethylene tubing-" O.D.