Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Acid Rain Science Fair Kit
GREENER_CHOICESCI_ED

Catalog No. S20640 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S20640 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer and reproductive harm.
Catalog No. S20640 Supplier Innovating Science™ Supplier No. IS5032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer and reproductive harm.

Ideal for use in the classroom or for science fairprojects to determine if you have acid rain in your area

  • Collect water samples and use a low range pH indicator with a colorimetric comparison chart.
  • Kit Includes:
    • 15 mL low range pH indicator
    • 6 test tubes
    • Color chart

Specifications

Color White
Grade 7 to 14
Age 13 to 20
Includes 15 mL Low Range pH Indicator, 6 Test tubes, Color Chart
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 1 Student
Product Type Acid Rain Science Fair Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.