Learn More
Ethos Biosciences Acetic Acid, Glacial, 1gal, Ethos
Catalog No. 5000574293
Encompass_Preferred
high-purity, concentrated form of acetic acid widely used in laboratory, industrial, and research applications. It serves as a key reagent in chemical synthesis, pH adjustment, buffer preparation, and histology staining procedures. Its anhydrous, concentrated formulation ensures consistent reactivity and reliable performance in both experimental and routine laboratory workflows. Produced under strict quality control standards, Glacial Acetic Acid is suitable for use in clinical, research, and educational laboratories, offering a versatile solution for applications that require a strong, consistent acid source while maintaining efficiency and reproducibility.
Catalog No. 50-005-74293 Supplier Ethos Biosciences Supplier No. 3298G
high-purity, concentrated form of acetic acid widely used in laboratory, industrial, and research applications. It serves as a key reagent in chemical synthesis, pH adjustment, buffer preparation, and histology staining procedures. Its anhydrous, concentrated formulation ensures consistent reactivity and reliable performance in both experimental and routine laboratory workflows. Produced under strict quality control standards, Glacial Acetic Acid is suitable for use in clinical, research, and educational laboratories, offering a versatile solution for applications that require a strong, consistent acid source while maintaining efficiency and reproducibility.